Tyvek Autoclave Bag with Steam Indicator
Leverandør: KEYSTONE CLEANROOM PRODUCTS
BGT2A2428
KACCBGT2A2428EA 38625 NOK
KACCBGT2A2428
Tyvek Autoclave Bag with Steam Indicator
Poser Steriliseringsposer
Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilisation of pharmaceutical change parts.
  • Tear and puncture resistant
  • Designed to be heat sealed closed
  • Superior to paper based autoclave pouches and wraps
  • Fast and consistent for equipment preparation area
  • Steam indicator and hydrophobic
  • Lint-free; strong factory seals
  • Exceptional microbial barrier
Standardise and simplify the way equipment is wrapped for steam sterilisation. The use of these bags will also save valuable time in theMore Product Information