Pro-Stand for Pro Line Micropipettes
Leverandør: KARTELL
0134800
KART0134800EA 3600 NOK
KART0134800
Pro-Stand for Pro Line Micropipettes
Pipettestativ
The sleek design and the rotating nature of this carousel stand allows the user to store or remove pipettes easily.
  • Stable base assists in single handed operation
  • Sturdy design ensures safe and secure pipette storage
  • Portable and lightweight design
This stand can hold both single channel and multichannel 'pro' micropipettes. It can be easily cleaned with soap and water, or common laboratory disinfectants.
The stand can accommodate the following Pro line pipettes: three multichannel micropipettes, 6 single channel micropipettes, or a combination ofMore Product Information