Stand for three pipettes
Leverandør: KARTELL
1315
KART1315EA 1900 NOK
KART1315
Stand for three pipettes
Pipettestativ
Transparent plastic holder for three Kartell micropipettes (Pluripet, Pluripet II, Fixopet, Labo!x) or pipettes from other brands.
  • Robust, easy to clean
  • Great stability thanks to the wide base
  • Sterilisable with radiation