Cuvettes for Olli-C-Analyzer®
Leverandør: KARTELL
2618
KART2618EA 6575 NOK
KART2618
Cuvettes for Olli-C-Analyzer®
Kyvetter Spesialkyvetter
These cylindrical cells are specifically used in conjunction with Olli-C-Analyzer.
  • Anti-shock polystyrene-moulding design
  • Highly resistant and dust-proof technology
  • Quality optical material suitable for IVD applications
Forpakning: In polystyrene dust proof boxes of 192 pieces.