Cuvettes for Fibrintimer®
Leverandør: KARTELL
2630
KART2630EA 1075 NOK
KART2630
Cuvettes for Fibrintimer®
Kyvetter Spesialkyvetter
Polystyrene cuvettes for Fibrintimer used to determine the coagulation-time of blood.
Sertifiseringer: IVD (in vitro diagnostic) certified.
Leveringsinformasjon: Supplied with 500 metallic mixers.