Soxhlet extractors
Leverandør: Kavalier
632442101100
KAVA632442101100EA 6075 NOK
KAVA632442101100
Soxhlet extractors
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
Borosilicate glass. For extraction from solids with solvents.
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.