Twisselmann extractors
Leverandør: Kavalier
632442108090
KAVA632442108090EA 76375 NOK
KAVA632442108090
Twisselmann extractors
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Complete assembly for 250 ml flask size.