Hanging balances, HCB and HCN Series
Leverandør: KERN SOHN
HCB20K10
KERNHCB20K10EA 3800 NOK
KERNHCB20K10
Hanging balances, HCB and HCN Series
Vekter
Ideal for rapid checking in goods receiving and shipping.
  • Data hold function: For easy reading of the weight, the display can be 'frozen' in different ways, either automatically when the weighing value remains unchanged or manually by pressing the Hold key
  • Peak load display (peak hold), measuring frequency 5 Hz
Sertifiseringer: Please note: If a calibration certificate is required, this should be requested when ordering the balance, as it can not be issued retrospectively.