Dewar flasks, Type MRD
Leverandør: KGW
11624
KGWA11624EA 19275 NOK
KGWA11624
Dewar flasks, Type MRD
Dewars og kryogene fartøy
Dewar flask is an insulated stirrer flask made up of borosilicate glass 3.3. It is used for storage of liquid or solid media's and solid matters coolants.
  • Compact design
  • Good tipping stability
  • Short distance to the stirring rod
  • Protective casing made of stucco aluminium
Suitable for mixing of substances in the range of −200 to +200 °C.
Area of application includes biotechnology, laboratory and medicinal technique.
Sertifiseringer: Conforms with DIN 12492, ISO 3585.