Cold trap, Type GKF
Leverandør: KGW
1702 1702-NW16 1702-NW25
KGWA1702EA 34175 NOK
KGWA1702 KGWA1702-NW16 KGWA1702-NW25
Cold trap, Type GKF
Vacuumfeller
This cold trap is a complete version in which the dewar flask is bonded to the cold trap, because of its design, this version has got two free walls for condensation. It is therefore particularly suited for trapping a high level of condensate from humidity, or solvents. This cold trap is equipped with a valve for draining off the condensate. It has got viewing strips as standard so that the LN₂ coolant level can easily be monitored. The standard cold trap is provided with conical ground joint connections NS 29/32.
  • Good chemical and thermal resistance
  • Viewing stripes for observation of