Blood collection tubes with KF+Na2EDTA
Leverandør: KIMA SAS DI CHIARIN RENZO
18506
KIMA18506EA 15125 NOK
KIMA18506
Blood collection tubes with KF+Na2EDTA
Diagnostiske systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PP, translucent.
  • KF+Na2 EDTA for 2,5ml of blood
  • Non sterile
  • With writing area