Blood collection tubes, K3EDTA, EMOPLASTIK®
Leverandør: KIMA SAS DI CHIARIN RENZO
18540125
KIMA18540125EA 0 NOK
KIMA18540125
Blood collection tubes, K3EDTA, EMOPLASTIK®
Diagnostiske systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PP, translucent, with cap.
  • K3 EDTA for 1 ml of blood
  • Non sterile
  • Label with 1 ml level mark
Blood collection tubes for pediatric low-volume drawing.