Wipes, KIMTECH PREP® SCOTTPURE™
Leverandør: KIMBERLY CLARK
61930
KIMB61930EA 6200 NOK
KIMB61930
Wipes, KIMTECH PREP® SCOTTPURE™
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
KIMTECH PREP® SCOTTPURE™ Wipers are designed for wiping tasks where precision and reliability are essential.
Low-lint rayon/polyester spunlace material for cloth-like softness. Engineered for excellent absorbency and low-lint wiping. Solvent resistant. No binders or surfactants. Applications include aerospace, clean manufacturing areas, DVD manufacturing, and printing.
Bestillingsinformasjon: Supplied without dispenser.