Wipe rolls, WypAll® L10 centrefeed food and hygiene
Leverandør: KIMBERLY CLARK
7255 7256
KIMB7255EA 1925 NOK
KIMB7255 KIMB7256
Wipe rolls, WypAll® L10 centrefeed food and hygiene
Papirservietter Tørkepapir og våtservietter Papirservietter til generell bruk
WypAll® L10 professional wiping paper products are designed for the specific environments where they will be used, helping enable you to deliver on your wiping task every time.
  • Single use wiping paper
  • Suitable for food contact
  • Centrefeed paper rolls are designed for use with centrefeed dispensers
  • WypAll® wiping paper is designed to be used once with detergent or disinfectant
Dry cleaning wipes designed for light duty hygiene wiping and perfect for tasks like cleaning food preparation and service surfaces. Embossing technology in the wiping paper givesMore Product Information