Disposable 12 mm generator, Polytron® PT-DA 12/2SYN-E123
Leverandør: KINEMATICA
11030035
KINE11030035EA 9525 NOK
KINE11030035
Disposable 12 mm generator, Polytron® PT-DA 12/2SYN-E123
Homogenisatorer Dispenseringverktøy
Disposable/reusable generators for when processing multiple samples per day, where the required sterilisation of stainless steel generators represents a significant time limitation.
  • Prevention of cross-contamination
  • Standard dispersing applications