Memo Rail Transmitters
Leverandør: KNICK
A1401N-P1-21
KNICA1401N-P1-21EA 11025 NOK
KNICA1401N-P1-21
Memo Rail Transmitters
Flytindikatorer
The Knick A1401N-P1-21, stands out in the market for its unparalleled performance and reliability.
  • Superior Quality
  • Cost-Effective
  • Fast Delivery
Memo Rail A1401N-P1-214-wire DIN for the determination of pH, Conductivity or dissolved oxygen with Memosens sensors measurement range can be selected via DIP switches2 with plug-P1screw.