pH electrode, SE 100 N
Leverandør: KNICK
SE100N
KNICSE100NEA 5975 NOK
KNICSE100N
pH electrode, SE 100 N
Elektroder pH-elektroder
This liquid-filled sensor is designed for tough daily use in the lab. The sensor is ideal for rapid sample change and low sample temperatures.
  • Integrated temperature sensor automatically compensates the pH measurement for temperature variations when the sensor is used in samples at different temperatures
  • Also fits in an Erlenmeyer flask