Sample bags, Whirl-Pak®
Leverandør: KOEHLER TECHNISCHE PRODUKTEN
1-7136
KOEH1-7136EA 1375 NOK
KOEH1-7136
Sample bags, Whirl-Pak®
Poser Prøveposer
Whirl-Pak plastic bags are made up of low density polyethylene (LDPE) which is suitable for liquid samples for food, water and waste water testing and also for biological or chemical samples.
  • Strong welding seam
  • Lodged with a metal strap for opening and closing of the bag
  • White inscription pad
  • Sterile bags with closure
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904