Cleaning swab
Leverandør: KOEHLER TECHNISCHE PRODUKTEN
2-1017
KOEH2-1017EA 170 NOK
KOEH2-1017
Cleaning swab
Svabere Svabere for rengjøring
These cotton swabs are designed for cleaning hardly accessible areas on optical devices and smears in bacteriology.