Pipette stand
Leverandør: KOEHLER TECHNISCHE PRODUKTEN
A-1650
KOEHA-1650EA 4050 NOK
KOEHA-1650
Pipette stand
Pipettestativ
Universal pipette stand made of rigid PVC. Accommodates up to 9 µl pipettes from various manufacturers.
  • Stable
  • Ergonomically tilted