Trypan blue

Leverandør: KOEHLER TECHNISCHE PRODUKTEN

Synonymer: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

CN76.2
KOEHCN76.2EA 1325 NOK
KOEHCN76.2
Trypan blue
Trypan blue
Formel: C₃₄H₂₄N₆Na₄O₁₄S₄
MDL nummer: MFCD00003969
CAS-nummer: 72-57-1
EINECS: 200-786-7

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR