Weighing dishes
Leverandør: Korff
60700
KORF60700EA 1850 NOK
KORF60700
Weighing dishes
Skip Veieskip
Dishes, made of aluminium.
  • Lightweight
  • Good temperature resistance
  • Good chemical resistance
Suitable for moisture determination. Thickness: 0,1 mm.