Modular assemblies for laboratory fume cupboards
Leverandør: KOETTERMANN
405-00028
KOTT405-00028EA 0 NOK
KOTT405-00028
Modular assemblies for laboratory fume cupboards
Møbler Kjemikalieskap
Modular fume cupboards are assembled from many different modules. The characteristic assembly for each type is called a 'variable assembly'.
  • One variable assembly is required for each modular fume cupboard
Fume cupboards are basically available with two different shipping options, which are part of the variable assemblies:
Knocked down: The front, sash, casing and support frame are shipped as assemblies and must be put together on site. As a rule, knocked-down fume cupboards of all types can be carried into laboratories even if the access is narrow.More Product Information