Penicillin : Streptomycin solution 0,06/0,1 g/L

Leverandør: Biowest

L0018-100
L0018-100EA 140 NOK
L0018-100
Penicillin : Streptomycin solution 0,06/0,1 g/L
Penicillin/Streptomycin

Penicillin-Streptomycin acts against gram-negative, gram-positive bacteria and mycoplasma.


  • pH of 6±1
  • Storage temperature of –20 °C
  • Shelf life of 24 months
  • Endotoxin: <10 EU/ml


NB: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Lagringstemperatur: Fryseskap
CAS-nummer: 1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR