1-(trans-Cinnamoyl)imidazole 98%

Leverandør: Thermo Scientific
Warning

L00441.06
L00441.06EA 505 NOK
L00441.06
1-(trans-Cinnamoyl)imidazole 98%
1-(trans-Cinnamoyl)imidazole

Purity: 98% CAS No.: 1138-15-4 Molecular Formula: C12H10N2O Formula Weight: 198.23 Storage Temperature: 2°C to 8°C Odour: Odourless Melting Point: 128-132° Beilstein Reference: 608846 MDL No.: MFCD00005288

Formel: C₁₂H₁₀N₂O
MW: 198,23 g/mol
Smeltepunkt: 128…132 °C
MDL nummer: MFCD00005288
CAS-nummer: 1138-15-4
EINECS: 214-510-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR