Cell culture media, Schneider's Insect Medium
Leverandør: Biowest
L0207-500
L0207-500EA 295 NOK
L0207-500
Cell culture media, Schneider's Insect Medium
Media, Cellekultur
Schneider’s Insect Medium was developed to support the growth of excised imaginal discs from the fruit fly, Drosophila melanogaster. When supplemented with 5 to 20% heat-inactivated foetal bovine serum Schneider's mediuam has been found to support the rapid growth of both primary and established cultures of cells derived from Drosophila spp. and several other dipterans.