Diphenyl phosphate 97

Leverandør: Thermo Fisher Scientific

Synonymer: Diphenyl hydrogen phosphate

L02121.06 L02121.14 L02121.22
L02121.06EA 630 NOK
L02121.06 L02121.14 L02121.22
Diphenyl phosphate 97
Diphenyl phosphate

Purity: 97% CAS No.: 838-85-7 Molecular Formula: C12H11O4P Formula Weight: 250.19 Melting Point: 62-70° Beilstein Reference: 1379164

Formel: C₁₂H₁₁O₄P
MW: 250.19 g/mol
Smeltepunkt: 62…70 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003033
CAS-nummer: 838-85-7
EINECS: 212-657-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR