Castor olje

Leverandør: Thermo Fisher Scientific

Synonymer: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

L04224.AK L04224.AU L04224.0F
L04224.AKEA 435 NOK
L04224.AK L04224.AU L04224.0F
Castor olje
Castor olje

CAS No.: 8001-79-4 Flash Point: 230°(446°F) Refractive Index: 1.4780 Boiling Point: 313° Merck Reference: 14,1898 Optical Rotation: +5° (c=5 in ethanol)

Kokepunkt: 313 °C
Smeltepunkt: –10 °C
Tetthet: 0,961
Flash punkt: 230 °C (446 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00130746
CAS-nummer: 8001-79-4
EINECS: 232-293-8
Merck Indeks: 14,1898

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR