Molekylsil 3A (0,3 nm, 3 Å), beads 1-2 mm (0.04-0.08 in)

Leverandør: Thermo Fisher Scientific

L05335.30 L05335.0B L05335.0I
L05335.30EA 830 NOK
L05335.30 L05335.0B L05335.0I
Molekylsil 3A (0,3 nm, 3 Å), beads 1-2 mm (0.04-0.08 in)
Molekylsil 3A (0,3 nm, 3 Å)

CAS No.: 308080-99-1 MDL No.: MFCD00131613

Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00131613
CAS-nummer: 308080-99-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR