Molekylsil 3A (0,3 nm, 3 Å), beads 3-5 mm (0.12-0.20 in)

Leverandør: Thermo Fisher Scientific

L05359.30 L05359.0B
L05359.30EA 795 NOK
L05359.30 L05359.0B
Molekylsil 3A (0,3 nm, 3 Å), beads 3-5 mm (0.12-0.20 in)
Molekylsil 3A (0,3 nm, 3 Å)

CAS No.: 308080-99-1 MDL No.: MFCD00131613

Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00131613
CAS-nummer: 308080-99-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR