O-(4-Nitrobenzyl)hydroxylamine hydrochloride 98%

Leverandør: Thermo Fisher Scientific

Synonymer: O-(4-Nitrobenzyl)-hydroxylammonium chloride

L05861.03 L05861.06
L05861.03EA 1150 NOK
L05861.03 L05861.06
O-(4-Nitrobenzyl)hydroxylamine hydrochloride 98%
O-(4-Nitrobenzyl)hydroxylamine hydrochloride

Purity: 98% CAS No.: 2086-26-2 Molecular Formula: C7H8N2O3.HCl Formula Weight: 204.61 Odour: Nearly Odourless Melting Point: ca 215° dec. Beilstein Reference: 3709565 MDL No.: MFCD00012954

Formel: C₇H₉ClN₂O₃
MW: 204.61 g/mol
Smeltepunkt: ∼ 215 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012954
CAS-nummer: 2086-26-2
EINECS: 218-228-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR