Versene
Leverandør: Biowest
L0630-100
L0630-100EA 230 NOK
L0630-100
Versene
Cellekultursupplementer og tilsetninger
Versene can be used instead of trypsin. It is a chelating agent that disperses the cells by cutting the cytoplasmic bridges between them. Versene has the advantage of being able to withstand autoclave temperature, which is a guarantee of sterility. In addition, it is less aggressive cells than trypsin, which is useful for studies on cell growth.