3-Methylstyrene ≥98% stabilised

Leverandør: Thermo Scientific
Danger

Synonymer: 3-Vinyltoluene

Stabilisering: stabilisert med 4-tert-Butylpyrokatekol 0,10 %

L08072.06
L08072.06EA 885 NOK
L08072.06
3-Methylstyrene ≥98% stabilised
3-Methylstyrene

Stabilized with 0.1% 4-tert-butylcatechol


Purity: 98% CAS No.: 100-80-1 Molecular Formula: C9H10 Formula Weight: 118.18 Storage Temperature: 2°C to 8°C Flash Point: 51°(123°F) Refractive Index: 1.5410 Odour: Pungent Melting Point: -82 to -81° Boiling Point: 170-171° Beilstein Reference: 1304618 UN No.: 2618 MDL No.: MFCD00008617

Formel: C₉H₁₀
MW: 118,18 g/mol
Kokepunkt: 170…171 °C
Smeltepunkt: –82 to –81 °C
Tetthet: 0,900
Flash punkt: 51 °C (123 °F)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008617
CAS-nummer: 100-80-1
EINECS: 202-889-2
FN: 2618
ADR: 3,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR