Phthalamide ≥97%

Leverandør: Thermo Fisher Scientific

L08600.09
L08600.09EA 590 NOK
L08600.09
Phthalamide ≥97%
Phthalamide

Purity: 97% CAS No.: 88-96-0 Molecular Formula: C8H8N2O2 Formula Weight: 164.16 Melting Point: 223° dec. Beilstein Reference: 1868220 Merck Reference: 14,7369 MDL No.: MFCD00025478

Formel: C₈H₈N₂O₂
Smeltepunkt: 223 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00025478
CAS-nummer: 88-96-0
EINECS: 201-870-6
Merck Indeks: 14,7369

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR