Cell detachment solution, Accutase
Leverandør: Biowest
L0950-100
L0950-100EA 1125 NOK
L0950-100
Cell detachment solution, Accutase
Cellekultursupplementer og tilsetninger
Accustase is a ready to use cell detachment solution of proteolytic and collagenolytic enzymes. Accutase is a direct gentle replacement for trypsin solution. It can also be used on suspension cells to reduce their agglutination in order to ease counting.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Arter
Kilde