1,3-Di-tert-butyltoluene ≥98%

Leverandør: Thermo Scientific

Synonymer: 1,3-Di-tert-butyl-5-methylbenzene

L11961.14
L11961.14EA 1525 NOK
L11961.14
1,3-Di-tert-butyltoluene ≥98%
1,3-Di-tert-butyltoluene

Purity: 98+% CAS No.: 15181-11-0 Molecular Formula: C15H24 Formula Weight: 204.36 Flash Point: 91°(195°F) Refractive Index: 1.4900 Melting Point: 31-32° Boiling Point: 243-244° Beilstein Reference: 2043032 MDL No.: MFCD00026300

Formel: C₁₅H₂₄
MW: 204,36 g/mol
Kokepunkt: 243…244 °C
Smeltepunkt: 31…32 °C
Tetthet: 0,860
Flash punkt: 91 °C (195 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00026300
CAS-nummer: 15181-11-0
EINECS: 239-230-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR