3,5-Dibromo-p-toluidine ≥99%

Leverandør: Thermo Fisher Scientific
Danger

L13101.06 L13101.03
L13101.06EA 2975 NOK
L13101.06 L13101.03
3,5-Dibromo-p-toluidine ≥99%
3,5-Dibromo-p-toluidine

Purity: 99% CAS No.: 13194-73-5 Molecular Formula: C7H7Br2N Formula Weight: 264.96 Melting Point: 90-92° Beilstein Reference: 2639285

Formel: C₇H₇Br₂N
Smeltepunkt: 90…92 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00151806
CAS-nummer: 13194-73-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR