2'-Deoxycytidine hydrochloride ≥99%

Supplier: Thermo Fisher Scientific

L14153.03 L14153.MD
L14153.03EA 895 NOK
L14153.03 L14153.MD
2'-Deoxycytidine hydrochloride ≥99%
2'-Deoxycytidine hydrochloride

Purity: 99% CAS No.: 3992-42-5 Molecular Formula: C9H13N3O4.HCl Formula Weight: 263.68 Odour: Odourless Melting Point: ca 170° dec. Beilstein Reference: 3576124 Optical Rotation: +57° (c=1 in water) MDL No.: MFCD00012840

Formula: C₉H₁₄ClN₃O₄
Melting Pt: ∼ 170 °C (dec.)
Storage Temperature: Romtemperatur
MDL Number: MFCD00012840
CAS Number: 3992-42-5
EINECS: 223-639-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR