Ethanolamine hydrochloride ≥98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 2-Aminoethanol hydrochloride, 2-Hydroxyethylammonium Chloride, Monoethanolamine Hydrochloride, 2-Aminoethan-1-ol hydrochloride

L14322.22 L14322.36
L14322.22EA 475 NOK
L14322.22 L14322.36
Ethanolamine hydrochloride ≥98%
Ethanolamine hydrochloride

Purity: 98+% CAS No.: 2002-24-6 Molecular Formula: HOCH2CH2NH2.HCl Formula Weight: 97.55 Melting Point: 83-86° Beilstein Reference: 3542892 Merck Reference: 14,3727 MDL No.: MFCD00012892

Formel: C₂H₈ClNO
MW: 97.54 g/mol
Smeltepunkt: 83…86 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012892
CAS-nummer: 2002-24-6
EINECS: 217-900-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR