Dodecamolybdofosforsyre hydrat (>47,5% Mo)

Leverandør: Thermo Fisher Scientific
Danger

L15827.14 L15827.22 L15827.36
L15827.14EA 515 NOK
L15827.14 L15827.22 L15827.36
Dodecamolybdofosforsyre hydrat (>47,5% Mo)
Dodecamolybdofosforsyre hydrat

CAS No.: 51429-74-4 Molecular Formula: H3PO4.12MoO3.xH2O Formula Weight: 1825.25(anhy) Melting Point: 78-90° Merck Reference: 14,7342 UN No.: 3085 MDL No.: MFCD00149913

Formel: H₃[P(Mo₃O₁₀)₄]·nH₂O
Smeltepunkt: 78…90 °C
Tetthet: 3,15
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149913
CAS-nummer: 51429-74-4
EINECS: 234-713-5
FN: 3085
ADR: 5.1,III
Merck Indeks: 14,7342

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR