1,1'-Dibromoferrocene ≥96%

Leverandør: Thermo Fisher Scientific
Warning

L17045.22
L17045.22EA 0 NOK
L17045.22
1,1'-Dibromoferrocene ≥96%
1,1'-Dibromoferrocene

Purity: 96% CAS No.: 1293-65-8 Molecular Formula: C10H8Br2Fe Formula Weight: 343.83 Storage Temperature: 2°C to 8°C Melting Point: 50-51° MDL No.: MFCD01075755

Formel: C₁₀Br₂Fe
Smeltepunkt: 50…51 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01075755
CAS-nummer: 1293-65-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR