4-Biphenylacetonitrile ≥97%

Leverandør: Thermo Fisher Scientific
Warning

L19243.04
L19243.04EA 1400 NOK
L19243.04
4-Biphenylacetonitrile ≥97%
4-Biphenylacetonitrile

Purity: 97% CAS No.: 31603-77-7 Molecular Formula: C14H11N Formula Weight: 193.25 Melting Point: 93-97° Boiling Point: 146-150°/0.03mm MDL No.: MFCD00016403

Formel: C₁₄H₁₁N
Kokepunkt: 146…150 °C (0,03 mmHg)
Smeltepunkt: 93…97 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016403
CAS-nummer: 31603-77-7

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR