Jars, wide mouth, with pressure cap
Leverandør: LAMAPLAST
2177 2184 2182 2178 2175 2174 2180
LAMA2177EA 35 NOK
LAMA2177 LAMA2184 LAMA2182 LAMA2178 LAMA2175 LAMA2174 LAMA2180
Jars, wide mouth, with pressure cap
Krukker Bokser, generell bruk
Jars, made of PS, crystal clear, transparent body with LDPE pressure cap.
Wide mouth universal containers for creams, semi solid substances, crystals, powders and granulates.