Jars, wide mouth, conical, graduated
Leverandør: LAMAPLAST
233 231 238C 238 235C 235 233C 231C
LAMA233EA 38 NOK
LAMA233 LAMA231 LAMA238C LAMA238 LAMA235C LAMA235 LAMA233C LAMA231C
Jars, wide mouth, conical, graduated
Krukker Bokser, generell bruk
PP, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Lid must be ordered separately
Wide mouth universal containers for creams, semi solid substances, crystals, powders and granulates.