Ultracool Circulation Chiller, UC 2
Leverandør: LAUDA
L003510
LAUDL003510EA 0 NOK
LAUDL003510
Ultracool Circulation Chiller, UC 2
Termostatbad Termostatbad, lukket Termostatbad med kjøling
Ultracool circulation chillers provide precise temperature control in an extended operating temperature range from −10 to +35 °C and a temperature stability of ±0,5 K.
  • Highly efficient industrial chiller
  • Graphic LCD with clear text menu navigation
  • Operation possible via web server
  • Electronic 2-point temperature control
  • Industrial block pump with high pressure and high flow rate
  • Inlet and outlet water connections in stainless steel
  • Integrated water filter, condenser cooling air
  • Operates with non flammable liquids (water,water/glycol)
The protectionMore Product Information