Biological safety cabinets, class II, type A2, Purifier® Cell Logic®+
Leverandør: LABCONCO
322691150
LBCP322691150EA 0 NOK
LBCP322691150
Biological safety cabinets, class II, type A2, Purifier® Cell Logic®+
Avtrekksskap og kabinetter Sikkerhetskabinetter
These Biosafety cabinets provide personnel, product and environmental protection from hazardous particulates such as agents that require Biosafety Level 1, 2 or 3 containment. Other appropriate applications include work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. All models are NSF compliant. Cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional canopy connection. When canopy-connected, the biosafety cabinets may be used forMore Product Information