Canopy connection kits for Purifier® Logic®+ biosafety cabinets
Leverandør: LABCONCO
3889201
LBCP3889201EA 0 NOK
LBCP3889201
Canopy connection kits for Purifier® Logic®+ biosafety cabinets
Avtrekksekstraktor
These kits are for thimble ducting class II, type A2 biosafety cabinets.
  • Overall height of the cabinet is 31,75 mm
Canopy provides an air gap to allow room air to enter the ductwork and mix with the filtered air being exhausted to the outside. Inadequate exhaust activates the switch in less than one second and triggers the cabinet to signal an audio and visual alarm via the LCD display. The motor also increases to maximum speed for personal safety during exhaust failure.