Ductless work stations, Protector®
Leverandør: LABCONCO
3930020 3930021
LBCP3930020EA 0 NOK
LBCP3930020 LBCP3930021
Ductless work stations, Protector®
Avtrekksskap og kabinetter Avtrekkshetter
Protector® work stations are specifically designed for the needs of pathologists and histotechnologists. Like a traditional fume hood, these enclosures have a front air foil and rear baffle to direct the inflow air across the work area and away from the operator. Ideal for cramped or crowded labs, protector work stations have a low profile so they fit easily under standard upper cabinets. Designed with a built-in blower which use specially treated carbon filters.
  • Face velocities from 60 to 100 fpm
  • Built-in dynamically-balanced motorized impeller
  • Ergonomic