Filtered PCR enclosures, Purifier®
Leverandør: LABCONCO
3970325
LBCP3970325EA 0 NOK
LBCP3970325
Filtered PCR enclosures, Purifier®
Avtrekksskap og kabinetter PCR-arbeidsstasjoner og -skap
Filtered PCR Enclosures direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination. These specialized vertical clean benches provide a controlled environment in which to perform polymerase chain reaction experiments. These enclosures include a variable digital timer and when the set time expires, the UV light automatically switches off in preparation for the next experiment.
  • Nominal downflow velocity of 45 to 65 fpm with aerodynamic air foil and disposible prefilter
  • UV-