Freeze dry systems using dry ice, 12 ports
Leverandør: LABCONCO
7522700
LBCP7522700EA 37575 NOK
LBCP7522700
Freeze dry systems using dry ice, 12 ports
Frysetørkere
Benchtop freeze drying system, type 304 stainless steel with 12 ports, -75 °C. Ideal for the laboratory with occasional lyophilisation requirements, the Dry Ice Benchtop Freeze Dry System provides a simple, economical means of freeze-drying. It is equipped with a center well for dry ice and solvent that serves as a water vapour collector and doubles as a convenient pre-freezing bath. Flasks, serum bottles and ampules may be frozen by dipping and rotating them in the well. Chamber size 248 x222 mm
  • Ideal for processing materials with low eutectic points
  • Includes dry ice/solvent center